Få mere at vide
T-2 Toxin, TRC
Artikelnummer. 30413585
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30413585 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413585 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0029805MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale T-2 Toxin (T-2)
CAS 21259-20-1
Molekylær formel C24 H34 O9
Mængde 5 mg
Synonym Trichothec-9-ene-3α,4β,8α,15-tetrol','12,13-epoxy-','4,15-diacetate 8-isovalerate (8CI), Spiro[2,5-methano-1-benzoxepin-10,2'-oxirane]','trichothec-9-ene-3,4,8,15-tetrol deriv., 4β,15-Diacetoxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-en-3α-ol, 8α-(3-Methylbutyryloxy)-4β,15-diacetoxyscirp-9-en-3α-ol, Fusariotoxin T 2, Insariotoxin, Mycotoxin T 2, NSC 138780, T 2, T 2 Toxin, T 2 mycotoxin, T-2 Lienomycin, Toxin T 2
InChI formel InChI=1S/C24H34O9/c1-12(2)7-18(27)32-16-9-23(10-29-14(4)25)17(8-13(16)3)33-21-19(28)20(31-15(5)26)22(23,6)24(21)11-30-24/h8,12,16-17,19-21,28H,7,9-11H2,1-6H3/t16-,17+,19+,20+,21+,22+,23+,24-/m0/s1
SMIL CC(C)CC(=O)O[C@H]1C[C@@]2(COC(=O)C)[C@H](O[C@@H]3[C@H](O)[C@@H](OC(=O)C)[C@@]2(C)[C@]34CO4)C=C1C
Molekylvægt (g/mol) 466.52
Formel vægt 466.2203