Learn More
IKA™ T 18 brushless digital Solution
Product Code. 31454106 Shop All IKA Products
Change view
Click to view available options
Plug Type:
CH
EU
UK
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type
31454106 CH
31454102 EU
31454095 UK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 31454106 Fournisseur IKA™ Code fournisseur 0010015151
  • ONLINE EXCLUSIVE PRICE
    22815.00 DKK
    21647.40 DKK / Pakke med 1
    Spare 1167.60 DKK
    5% Slukket
Afficher toutes les promotions
 Restricted Item
På lager
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
The T 18 brushless digital Solution offers a complete starter setup for homogenization, including a dispersing tool, shaft, and stand. Ideal for industries operating under ambient conditions requiring high hygiene and regulatory compliance.
The T 18 brushless digital is an economical high performance dispersing instrument with a wide speed range of 3000 - 25 000 rpm for working volumes of up to 1.500 mL (H2O). It enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantee a wide range of applications for efficient emulsification, homogenization, dispersing and suspending and a reduction of particle sizes to the micrometer range. With the following features, this disperser meets the needs of a modern laboratory:
  • Brushless motor for improved motor life, quieter operation and hygienic applications
  • Digital speed display and setting
  • Timer function for unattended operation
  • Quick coupling for easy and fast exchange of dispersing elements for cleaning and maintenance
  • USB interface for remote control and parameter documentation with the laboratory software labworldsoft™
  • Wide range of dispersing tools available; stainless steel with high chemical resistance or disposable plastic tools
The T 18 brushless digital Solution provides a complete starter solution to start your homogenisation journey, complete with a dispersing tool, shaft and stand, ideal for applications in industries working in ambient conditions that require a high degree of hygience and compliance.
TRUSTED_SUSTAINABILITY
Spécification
Product Type Homogenizer Package
Speed ≤460 rpm
Voltage 220/240 V
Packaging Type Individual Pack
Plug Type CH
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.