Learn More
CELLPATH SYSTEM II HEX-CASSETTE LID - WHITE (LOOSE)
Product Code. 30226232
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 1000
Product Code. Color Unit Size
30226232 White Pack of 1000
1 options
Product Code. 30226232 Supplier CELLPATH Supplier No. EAE010203A
  • ONLINE EXCLUSIVE PRICE
    562.49 DKK
    266.19 DKK / Pakke med 1000
    Spare 296.30 DKK
    52% Slukket
 Restricted Item
Forventet forsendelse: 22-12-2026
The System II Hex Cassette Lid is compatible with the System II Hex Cassette. Users can choose an easy to attach disposable plastic lid, or a re-useable stainless-steel version depending on their requirements.
Specifications
Color White
Product Type Cassette