Learn More
CELLPATH SYSTEM II HEX - CASSETTE
Product Code. 30226228
Change view
Click to view available options
Material:
Laser Threaded
Loose
Unit Size:
Pack of 1000
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226228 Loose Pack of 1000
30226230 - Pack of 2000
30226231 Laser Threaded Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226228 Supplier CELLPATH Supplier No. EAD010203A
  • ONLINE EXCLUSIVE PRICE
    608.37 DKK
    287.85 DKK / Pakke med 1000
    Spare 320.52 DKK
    52% Slukket
 Restricted Item
Forventet forsendelse: 09-12-2026
The innovative honeycomb design of the The System II Hex Cassette maximises the open surface area compared to traditional round perforations, increasing reagent flow through for improved processing and a more rigid block when sectioning.
  • The System II Hex Cassette benefits from an optimised writing surface as well as extra thick side walls that increase stability and rigidity, essential for high-quality sectioning. Supplied loose, threaded (for easy loading of Leica and Sakura printers) or pre-loaded in hopper tubes (for easy loading of Thermo Scientific cassette printers).
  • Available in a range of colours, the System II Hex Cassette can be closed by using one of our plastic or stainless-steel lids.
  • Key Benefits:
  • Hex design maximises open surface area to increase the fluid exchange
  • Optimised writing surface for printing high-quality text, 1D and 2D barcodes
  • Writing surface suitable for marking with a CellMark marker pen and CellMark 2B pencil
  • Large colour range available
Specifications
Color White
Material Loose
Product Type Cassette