Learn More
CELLPATH SYSTEM II+ 4 WELL CASSETTE
Product Code. 30226261
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226261 Threaded Pack of 2000
30226259 Loose Pack of 1000
30226260 Hoppered Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226261 Supplier CELLPATH Supplier No. EAM180272B
  • ONLINE EXCLUSIVE PRICE
    4138.14 DKK
    1957.33 DKK / Pakke med 2000
    Spare 2180.81 DKK
    52% Slukket
 Restricted Item
Forventet forsendelse: 15-12-2026
The System II+ 4 Well Cassette incorporates multiple wells into the cassette allows multiple biopsies to be processed simultaneously, significantly reducing user workload. They come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Threaded
Product Type Cassette