Få mere at vide
Syrosingopine, TRC
Artikelnummer. 30426088
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426088 5 mg
30423594 50 mg
30411808 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426088 Leverandør Toronto Research Chemicals Leverandørnr. TRCS9200685MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 84-36-6
Molekylær formel C35 H42 N2 O11
Molekylvægt (g/mol) 666.71
Synonym Yohimban-16-carboxylic acid, 18-[[4-[(ethoxycarbonyl)oxy]-3,5-dimethoxybenzoyl]oxy]-11,17-dimethoxy-, methyl ester, (3β,16β,17α,18β,20α)-, 3β,20α-Yohimban-16β-carboxylic acid, 18β-hydroxy-11,17α-dimethoxy-, methyl ester, 4-hydroxy-3,5-dimethoxybenzoate (ester) ethyl carbonate (ester) (8CI), Reserpic acid, methyl ester, ester with syringic acid, ethyl carbonate (6CI), Carbethoxysyringoyl methylreserpate, Isotense, Londomin, Methyl carbethoxysyringoyl reserpate, Methyl reserpate ester of syringic acid ethyl carbonate, NSC 77030, Raunova, Seniramin, Singoserp, Siringina, Su 3118, Syringic acid ethyl carbonate ester with methyl reserpate, Syringopine, Syrosingopin, Syrosingopine
IUPAC navn methyl (1R,15S,17R,18R,19S,20S)-17-(4-ethoxycarbonyloxy-3,5-dimethoxybenzoyl)oxy-6,18-dimethoxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMIL CCOC(=O)Oc1c(OC)cc(cc1OC)C(=O)O[C@@H]2C[C@@H]3CN4CCc5c([nH]c6cc(OC)ccc56)[C@H]4C[C@@H]3[C@@H]([C@H]2OC)C(=O)OC
Tekniske data
Kemisk navn eller materiale Syrosingopine
Mængde 5 mg
InChI formel InChI=1S/C35H42N2O11/c1-7-46-35(40)48-31-26(42-3)12-18(13-27(31)43-4)33(38)47-28-14-19-17-37-11-10-22-21-9-8-20(41-2)15-24(21)36-30(22)25(37)16-23(19)29(32(28)44-5)34(39)45-6/h8-9,12-13,15,19,23,25,28-29,32,36H,7,10-11,14,16-17H2,1-6H3/t19-,23+,25-,28-,29+,32+/m1/s1
Formel vægt 666.2789