Få mere at vide
Syringic-d6 Acid, TRC
Artikelnummer. 30416603
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30416603 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416603 Leverandør Toronto Research Chemicals Leverandørnr. TRCS92002710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Syringic-d6 Acid
CAS 84759-06-8
Molekylær formel C9 D6 H4 O5
Mængde 10 mg
Synonym Benzoic acid','4-hydroxy-3,5-di(methoxy-d3)- (9CI), 3,5-Dimethoxy-d6-4-hydroxybenzoic Acid, Syringic-d6 Acid, Syringic Acid-D6 (dimethoxy-D6), 2,6-Di(methoxy-d3)-4-carboxyphenol, 3,5-Di(methoxy-d3)-4-hydroxybenzoic acid, Gallic acid 3,5-di(methyl-d3) ether
InChI formel InChI=1S/C9H10O5/c1-13-6-3-5(9(11)12)4-7(14-2)8(6)10/h3-4,10H,1-2H3,(H,11,12)/i1D3,2D3
SMIL [2H]C([2H])([2H])Oc1cc(cc(OC([2H])([2H])[2H])c1O)C(=O)O
IUPAC navn 4-hydroxy-3,5-bis(trideuteriomethoxy)benzoic acid
Molekylvægt (g/mol) 204.2097
Formel vægt 204.0905