Learn More
Trajan Scientific and Medical™ Syringe
For reduced non specific sample interaction and increased analyte recovery
Brand:  Trajan Scientific and Medical™ 000440
Additional Details : Weight : 0.10000kg
Product Code. 10781422
  • 1115.00 DKK / Each
Estimated Shipment: 03-07-2024
to see stock.
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Shimadzu instruments
Specifications
Specifications
26 G
0.47 mm
1/Ea.
Cone
0.155 mm
Syringe
0.5 μL
Shimadzu