Learn More
Syngene™ UV Transilluminator 2020M
Product Code. 12874028 Shop All Syngene Products
Change view
Click to view available options
Wavelength:
302 nm
Unit Size:
Each
Product Code. Wavelength Unit Size
12874028 302 nm Each
1 options
Product Code. 12874028 Supplier Syngene™ Supplier No. BT2020ME
  • ONLINE EXCLUSIVE PRICE
    8360.00 DKK
    7848.67 DKK / Pakke med 1
    Spare 511.33 DKK
    6% Slukket
View all promotions
Forventet forsendelse: 26-01-2027
UV Transilluminator 2020M is for illumination of any sample excited by UV light.
  • Sealed to allow wet gels on surface
  • For use in Syngene Systems
Specifications
Product Type Single UV Transilluminator
Voltage 100/115/220/230 V
Intensity Fixed
Wavelength 302 nm
Lighting Type UV