Learn More
OHAUS™ Swivel Multi-Purpose Clamps
OHAUS™ Swivel Multi-Purpose Clamps are designed to securely grip and position a variety of laboratory glassware.
Brand: OHAUS™ 30392226
814.68 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in nickel-plated zinc or stainless steel
- Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifications
Multi-purpose Clamp | |
Non-slip vinyl sleeves and fiberglass covers | |
6.42 in. | |
Medium | |
2-Prong Single Adjust |
Stainless Steel | |
Clamp, Multi Purpose, CLM-SWIVL2SSM | |
163.00 mm | |
0 to 76 mm |
Product Content Correction
Product Title
OHAUS™ Swivel Multi-Purpose Clamps > Model: CLM-SWIVL2SSM
Spot an opportunity for improvement?Share a Content Correction