Learn More
Pyrex™ SVL™ Stirrer Heads for SVL™ Stirrer Shafts
Product Code. 12024422
Change view
Click to view available options
Type:
Anchor
Collapsible Impeller
Helix - 6 Blades
Paddle
Diameter (Metric):
54 mm
70 mm
80 mm
90 mm
Unit Size:
Each
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Diameter (Metric) Unit Size
12024422 Collapsible Impeller 70 mm Each
12044422 Anchor 90 mm Each
12054422 Helix - 6 Blades 80 mm Each
15527740 Paddle 70 mm Pack of 5
15537740 Anchor 54 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12024422 Supplier Pyrex™ Supplier No. 58893
  • ONLINE EXCLUSIVE PRICE
    670.00 DKK
    493.69 DKK / Pakke med 1
    Spare 176.31 DKK
    26% Slukket
Forventet forsendelse: 11-01-2027
Switch stirring heads quickly and easily. Pyrex™ SVL™ Stirrer Head for SVL™ Stirrer Shafts are constructed of durable PTFE polymer; some types are fortified with a metal core.
  • Designed for use with SVL™ stirrer shafts
  • Head simply screws into the end of the shaft
  • Manufactured from a PTFE based polymer with the Anchor and Helix types stiffened by a metal core
Specifications
Diameter (Metric) 70 mm
For Use With (Equipment) Vessels with SVL 30 joint or 29/32 socket
Quantity 1 Each
Type Collapsible Impeller