missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ SV2A Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 11386553
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
Pakningsstørrelse:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11386553 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11386553 Supplier Invitrogen™ Supplier No. PA518581

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, human, mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

This protein plays a role in the control of regulated secretion in neural and endocrine cells, enhancing selectively low-frequency neurotransmission. Positively regulates vesicle fusion by maintaining the readily releasable pool of secretory vesicles.It is identified as the brain binding-site for the antiepileptic drug levetiracetam/lev.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SV2A
Applikationer Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SV2A
Genadgangsnr. Q02563, Q7L0J3
Gene Alias AI746429; calcium regulator SV2A; Kiaa0736; mKIAA0736; PSEC0174; SV2; SV2A; synaptic vesicle glycoprotein 2 a; synaptic vesicle glycoprotein 2a; Synaptic vesicle protein 2; synaptic vesicle protein 2A; synaptic vesicle transmembrane transporter (SV2) RNA
Gensymboler SV2A
Værtsarter Goat
Immunogen Synthetic peptide sequence (TVELYPSDKRTTA) corresponding to the internal amino acids of SV2A (aa 673-685).
Oprensningsmetode Ammonium sulfate precipitation
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 117559, 9900
Målarter Human, Rat
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.