Learn More
Tork Surface Cleaning Wet Wipe Bucket W15
Product Code. 30243699 Shop All Tork Products
Change view
Click to view available options
Includes:
Bucket
Refill Only
Unit Size:
4 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
30243699 Refill Only 4 rolls
30243825 Bucket 4 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243699 Supplier Tork Supplier No. 190694
  • ONLINE EXCLUSIVE PRICE
    886.08 DKK
    813.70 DKK / 4 ruller
    Spare 72.38 DKK
    8% Slukket
Forventet forsendelse: 15-12-2026
The Tork Surface Cleaning Wet Wipes are convenient and efficient for surface cleaning when water is not available, while it protects sensitive surfaces, electronics and processes.
  • Clean surfaces effectively with these professional-quality wet wipes.
  • Remove oil, grease, lubricants and dirt with ease, thanks to their strong texture.
  • Maintain surface hygiene levels when water isn't available with this convenient design.
  • Protect sensitive surfaces and electronics while keeping them clean.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wet Wipe
Length (English) 17.13 yd.
Length (Metric) 15.66 m
Width (English) 5.31 in.
Width (Metric) 13.5 cm
Certifications/Compliance FSC MIX Credit
Color White
Ply 1-ply
Includes Refill Only
Certifications/Compliance (Europe) FSC MIX Credit