Learn More
CELLPATH SUPA MEGA SLIM CASSETTE - WHITE
Product Code. 30226266
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
Product Code. Color Unit Size
30226266 White Pack of 100
1 options
Product Code. 30226266 Supplier CELLPATH Supplier No. EAN010202A
  • ONLINE EXCLUSIVE PRICE
    1595.99 DKK
    754.88 DKK / Pakke med 100
    Spare 841.11 DKK
    52% Slukket
 Restricted Item
Forventet forsendelse: 15-12-2026
The Supa Mega Slim is being widely adopted in clinical, research and toxicology laboratories. Its internal chamber is one-third of the depth of the conventional Supa Mega Cassette.
  • CAP in the USA2.3, through reducing specimen thickness and shortening the processing time. Through using the Slim, casting can be done in only 20 minutes at -8°C.
  • Key Benefits:
  • Better reagent flow and improved turnaround times
  • Casting can be done in 20 minutes at -8°C
  • Less wax used during the embedding process
  • Large colour range available
  • Specific base moulds
Specifications
Color White
Product Type Cassette