missing translation for 'onlineSavingsMsg'
Få mere at vide

Sunset Yellow (E110) D4 (phenyl D4), TRC

Artikelnummer. 30427480
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427480 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427480 Leverandør Toronto Research Chemicals Leverandørnr. TRCS82500550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Sunset Yellow (E110) D4 (phenyl D4)
CAS 2259674-84-3
Molekylær formel C16 D4 H6 N2 O7 S2 . 2 Na
Mængde 50 mg
Synonym Sunset Yellow (E110) D4 (Phenyl D4), 6-Hydroxy-5-(4-sulfophenylazo)-2-naphthalenesulfonic acid D4 (4-sulfophenyl-2,3,5,6-D4) disodium salt, 6-Hydroxy-5-(4-sulfophenylazo-2,3,5,6-d4)-2-naphthalenesulfonic acid disodium salt
InChI formel InChI=1S/C16H12N2O7S2.2Na/c19-15-8-1-10-9-13(27(23,24)25)6-7-14(10)16(15)18-17-11-2-4-12(5-3-11)26(20,21)22;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2/i2D,3D,4D,5D;;
SMIL [Na+].[Na+].[2H]c1c([2H])c(c([2H])c([2H])c1N=Nc2c(O)ccc3cc(ccc23)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC navn disodium;6-hydroxy-5-[(E)-(2,3,5,6-tetradeuterio-4-sulfonatophenyl)diazenyl]naphthalene-2-sulfonate
Molekylvægt (g/mol) 456.394
Formel vægt 455.9976
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.