Få mere at vide
Sunset Yellow (E110) D4 (phenyl D4), TRC
Artikelnummer. 30415535
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30415535 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415535 Leverandør Toronto Research Chemicals Leverandørnr. TRCS825005100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sunset Yellow (E110) D4 (phenyl D4)
CAS 2259674-84-3
Molekylær formel C16 D4 H6 N2 O7 S2 . 2 Na
Mængde 100 mg
Synonym Sunset Yellow (E110) D4 (Phenyl D4), 6-Hydroxy-5-(4-sulfophenylazo)-2-naphthalenesulfonic acid D4 (4-sulfophenyl-2,3,5,6-D4) disodium salt, 6-Hydroxy-5-(4-sulfophenylazo-2,3,5,6-d4)-2-naphthalenesulfonic acid disodium salt
InChI formel InChI=1S/C16H12N2O7S2.2Na/c19-15-8-1-10-9-13(27(23,24)25)6-7-14(10)16(15)18-17-11-2-4-12(5-3-11)26(20,21)22;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2/i2D,3D,4D,5D;;
SMIL [Na+].[Na+].[2H]c1c([2H])c(c([2H])c([2H])c1N=Nc2c(O)ccc3cc(ccc23)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC navn disodium;6-hydroxy-5-[(E)-(2,3,5,6-tetradeuterio-4-sulfonatophenyl)diazenyl]naphthalene-2-sulfonate
Molekylvægt (g/mol) 456.394
Formel vægt 455.9976