missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ Sunitinib malate
GREENER_CHOICE

Artikelnummer. 18792544
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18792544 10 mg
18772724 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 18792544 Leverandør Tocris Bioscience™ Leverandørnr. 3768/10

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy

Potent VEGFR, PDGFRβ and KIT inhibitor

Potent, ATP-competitive VEGFR, PDGFRβ and KIT inhibitor (Ki values are 2, 9, 17, 8 and 4 nM for VEGFR -1, -2, -3, PDGFRβ and KIT respectively). Also inhibits cellular receptor phosphorylation of FLT3, RET and CSF-1R. Exhibits antiangiogenic and antitumor activity in multiple xenograft models.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341031-54-7
Molekylær formel C26H33FN4O7

Specifications

Kemisk navn eller materiale Sunitinib malate
Mængde 10 mg
Formel vægt 532.56
Renhed >99%
Molekylvægt (g/mol) 532.569
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.