Få mere at vide
Sumatriptan N-Oxide, TRC
Artikelnummer. 30138685
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138685 50 mg
30135509 25 mg
17983755 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138685 Leverandør Toronto Research Chemicals Leverandørnr. TRCS81001050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 212069-94-8
Molekylær formel C14 H21 N3 O3 S
Molekylvægt (g/mol) 311.4
Synonym 3-[2-(Dimethyloxidoamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide, GR 112504, Sumatriptan N-Oxide, GR 112504X, Ph Eur Sumatriptan Impurity D, BP Sumatriptan Impurity D, Sumatriptan N-oxide (GR 112504X), Ph Eur Sumatriptan Impurity Mixture, Sumatriptan Test Mix for Impurity Identification Solution 2
IUPAC navn N,N-dimethyl-2-[5-(methylsulfamoylmethyl)-1H-indol-3-yl]ethanamine oxide
SMIL CNS(=O)(=O)Cc1ccc2[nH]cc(CC[N+](C)(C)[O-])c2c1
Tekniske data
Kemisk navn eller materiale Sumatriptan N-Oxide
Mængde 50 mg
InChI formel InChI=1S/C14H21N3O3S/c1-15-21(19,20)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2,3)18/h4-5,8-9,15-16H,6-7,10H2,1-3H3
Formel vægt 311.1304