Få mere at vide
Sulthiame-d4, TRC
Artikelnummer. 30422086
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30422086 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422086 Leverandør Toronto Research Chemicals Leverandørnr. TRCS78950210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulthiame-d4
CAS 1795021-05-4
Renhedsgrad noter HPLC
Molekylær formel C10H10D4N2O4S2
Mængde 10 mg
Synonym 4-(Tetrahydro-1,1-dioxido-2H-1,2-thiazin-2-yl)benzenesulfonamide-d4, 4-(Tetrahydro-2H-1,2-thiazin-2-yl)benzenesulfonamide-d4 S,S-Dioxide, 2-(p-Sulfamoylphenyl)-tetrahydro-2H-1,2-thiazine-d4 1,1-Dioxide, Bayer A 168-d4, Conadil-d4, Contravul-d4, Elisal-d4, Ospolot-d4, Riker 594-d4, Sulphenyltame-d4, Sulphenytame-d4, Sulphthiame-d4, Sulthiam-d4, Sulthiamine-d4, Sultiam-d4, Sultiame-d4, Trolone-d4, p-(Tetrahydro-2H-1,2-thiazin-2-yl)benzenesulfonamide-d4 Dioxide
InChI formel InChI=1S/C10H14N2O4S2/c11-18(15,16)10-5-3-9(4-6-10)12-7-1-2-8-17(12,13)14/h3-6H,1-2,7-8H2,(H2,11,15,16)/i3D,4D,5D,6D
SMIL NS(C1=C([2H])C([2H])=C(N2S(CCCC2)(=O)=O)C([2H])=C1[2H])(=O)=O
IUPAC navn 2,3,5,6-tetradeuterio-4-(1,1-dioxothiazinan-2-yl)benzenesulfonamide
Molekylvægt (g/mol) 294.38
Formel vægt 294.0646
Procent renhed >95