missing translation for 'onlineSavingsMsg'
Learn More

Sultamicillin Tosylate, TRC

Product Code. 30427550
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30427550 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427550 Supplier Toronto Research Chemicals Supplier No. TRCS789490500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Sultamicillin tosilate
CAS 83105-70-8
Molekylær formel C25 H30 N4 O9 S2 . C7 H8 O3 S
Mængde 500 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-','[[[(2S,6R)-3,3-dimethyl-4,4-dioxido-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]oxy]methyl ester','(2S,5R,6R)-','(4-methylbenzenesulfonate) (1:1), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[(aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-','[[(3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl)carbonyl]oxy]methyl ester','S,S-dioxide','[2S-[2α(2R*,5S*),5α,6β(S*)]]-','mono(4-methylbenzenesulfonate), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-aminophenylacetyl]amino]-3,3-dimethyl-7-oxo-','[[[(2S,6R)-3,3-dimethyl-4,4-dioxido-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl]carbonyl]oxy]methyl ester','(2S,5R,6R)-','mono(4-methylbenzenesulfonate) (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[(aminophenylacetyl)amino]-3,3-dimethyl-7-oxo-','[[(3,3-dimethyl-4,4-dioxido-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-yl)carbonyl]oxy]methyl ester','[2S-[2α(2R*,5S*),5α,6β(S*)]]-','mono(4-methylbenzenesulfonate), Bacimex, Bethacil orale, Sultamicillin p-toluenesulfonate, Sultamicillin tosylate, Unacid PD oral, Unacim orale
InChI formel InChI=1S/C25H30N4O9S2.C7H8O3S/c1-24(2)17(29-20(32)16(21(29)39-24)27-19(31)15(26)12-8-6-5-7-9-12)22(33)37-11-38-23(34)18-25(3,4)40(35,36)14-10-13(30)28(14)18;1-6-2-4-7(5-3-6)11(8,9)10/h5-9,14-18,21H,10-11,26H2,1-4H3,(H,27,31);2-5H,1H3,(H,8,9,10)/t14-,15-,16-,17+,18+,21-;/m1./s1
SMIL Cc1ccc(cc1)S(=O)(=O)O.CC2(C)S[C@@H]3[C@H](NC(=O)[C@H](N)c4ccccc4)C(=O)N3[C@H]2C(=O)OCOC(=O)[C@@H]5N6[C@@H](CC6=O)S(=O)(=O)C5(C)C
IUPAC navn [(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carbonyl]oxymethyl (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;4-methylbenzenesulfonic acid
Molekylvægt (g/mol) 766.86
Formel vægt 766.1648
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.