missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | SULT1A4 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828413
|
Proteintech
11376-1-AP-20UL |
20 ÎĽL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16818413
|
Proteintech
11376-1-AP-150UL |
150 ÎĽL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SULT1A4 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P0DMN0 | |
445329 | |
SULT1A4 Fusion Protein Ag1935 | |
Primary | |
-20°C | |
SULT1A4 |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Aryl sulfotransferase 1A3/1A4, HAST3, M PST, ST1A3/ST1A4, STM, Sulfotransferase 1A3/1A4, SULT1A3, SULT1A4, TL PST | |
SULT1A4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SULT1A4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech