Få mere at vide
Sulpiride EP Impurity G, Mikromol™
Artikelnummer. 30459345
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459345 4 x 25 mg
30049412 25 mg
30050018 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459345 Leverandør Mikromol Leverandørnr. MM0051.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sulpiride EP Impurity G, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-Desmethylsulpiride
CAS 67381-52-6
Analyt- eller komponentnavne O-Desmethylsulpiride
Forsendelsestilstand Room Temperature
Molekylær formel C14 H21 N3 O4 S
Certificeringer/compliance ISO 17025
Synonym (RS)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulphamoylbenzamide, Sulpiride Imp. G (EP), O-Desmethylsulpiride, Sulpiride Impurity G
InChI formel InChI=1S/C14H21N3O4S/c1-2-17-7-3-4-10(17)9-16-14(19)12-8-11(22(15,20)21)5-6-13(12)18/h5-6,8,10,18H,2-4,7,9H2,1H3,(H,16,19)(H2,15,20,21)
Løsningstype Neat
SMIL CCN1CCCC1CNC(=O)c2cc(ccc2O)S(=O)(=O)N
IUPAC navn N-[(1-ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulfamoylbenzamide
Molekylvægt (g/mol) 327.4
Mængde 4 x 25 mg
Formel vægt 327.1253 g/mol