missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.005M, Chem-Lab

Catalog Number p-7114737
Click to view available options
Quantity:
5 L
25 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

For laboratory use, Titrations and TMAR Sulfates determination, ASTM D-3228

Concentration 0.491g H2SO4 / 1L H2O
Density 1.00g/mL
Packaging PE
Normality 0.01N (±0.00002 at 20°C)
Quantity 5 L
Formula Weight 98.08g/mol
Chemical Name or Material Sulfuric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'