missing translation for 'onlineSavingsMsg'
Learn More

Sulfur, Reagent grade, Purified by refining, Honeywell™

Catalog Number p-7120848
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]
This item is not returnable. View return policy
7704-34-9
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
5362487
CHEBI:26833
sulfur
[S]

Reagent Grade, powder, purified by refining, -100 mesh particle size

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Point 445°C
Melting Point 120°C
Color Yellow
Physical Form Powder
Quantity 500 g
UN Number UN1350
Flash Point 207°C (404.6°F)
Formula Weight 32.07g/mol
Grade Reagent
Ignition Point 450°F
Packaging Poly Bottle
Vapor Pressure 1mmHg (183.8°C), 10mmHg (246°C)
Density 2.07g/cu.cm; Vapor Density: 8.9 (vs air)
Chemical Name or Material Sulfur, Purified by refining
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'