Få mere at vide
Sulfur in heavy mineral oil, ASTM D2622, Calibration Set C: 1.0000 % (10000ppm), 2.0000 % (20000ppm), 3.0000 % (30000ppm), 4.0000 % (40000ppm), 5.0000 % (50000ppm) . 100 mL ea., Paragon Scientific™ Artikelnummer.: 16020202
Sulfur in heavy mineral oil, ASTM D2622, Calibration Set C: 1.0000 % (10000ppm), 2.0000 % (20000ppm), 3.0000 % (30000ppm), 4.0000 % (40000ppm), 5.0000 % (50000ppm) . 100 mL ea., Paragon Scientific™
Artikelnummer. 16020202
Set of 5 = x5 100mLs, 5ml
Mængde:
Set of 5 = x5 100mLs
Pakningsstørrelse:
5ml
Artikelnummer. 16020202
Brand: Paragon Scientific D2622HMSUC
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Se alternative produkter
Suitable for calibration, verification or for use in quality control procedures used for analysing sulfur content in petroleum products, with sulfur content ranging from 0 (Blank) through to 5.0000% (50000ppm) for each material.
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation.
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide.
  • Suitable for but not limited to internationally recognised test methods and protocol: ASTM D2622, ASTM D4294, ASTM D5453, ASTM D7039, IP 336, IP 496, IP 497.
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data.
  • Supplied in 100 mL volume, tamper-evident glass packaging, providing assurance of sample integrity.
  • Manufactured in the United Kingdom.
  • 12 month shelf life.
Tekniske data
Mængde Set of 5 = x5 100mLs
Nominal values quoted. Certified Values will vary from batch to batch.
Korrektion af produktindhold
Produkttitel
Sulfur in heavy mineral oil, ASTM D2622, Calibration Set C: 1.0000 % (10000ppm), 2.0000 % (20000ppm), 3.0000 % (30000ppm), 4.0000 % (40000ppm), 5.0000 % (50000ppm) . 100 mL ea., Paragon Scientific™ >