Få mere at vide
Sulforaphene, TRC
Artikelnummer. 30133905
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133905 100 mg
17960625 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133905 Leverandør Toronto Research Chemicals Leverandørnr. TRCS699440100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 592-95-0
Molekylær formel C6 H9 N O S2
Molekylvægt (g/mol) 175.27
Synonym Raphanin, Sulforaphen, (-)-Sulforaphene, 4-Isothiocyanato-1-(methylsulfinyl)-1-butene
IUPAC navn (E)-4-isothiocyanato-1-methylsulfinyl-but-1-ene
SMIL CS(=O)\C=C\CCN=C=S
Tekniske data
Kemisk navn eller materiale L-Sulforaphene
Mængde 100 mg
InChI formel InChI=1S/C6H9NOS2/c1-10(8)5-3-2-4-7-6-9/h3,5H,2,4H2,1H3/b5-3+
Formel vægt 175.0126