Få mere at vide
Sulfo-N-succinimidyl 6-[3-(2-Pyridyldithio)propionamido] Hexanoate, Sodium Salt (Technical Grade), TRC
Artikelnummer. 30411351
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411351 50 mg
30422684 5 mg
30422706 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30411351 Leverandør Toronto Research Chemicals Leverandørnr. TRCS75500350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 169751-10-4
Molekylær formel C18 H22 N3 O8 S3 . Na
Molekylvægt (g/mol) 527.57
Synonym 2,5-Dioxo-1-[[1-oxo-6-[[1-oxo-3-(2-pyridinyldithio)propyl]amino]hexyl]oxy]-3-pyrrolidinesulfonic Acid Monosodium Salt, Sulfo-LC-SPDP
IUPAC navn sodium;2,5-dioxo-1-[6-[3-(pyridin-2-yldisulfanyl)propanoylamino]hexanoyloxy]pyrrolidine-3-sulfonate
SMIL [Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)CCCCCNC(=O)CCSSc2ccccn2)C1=O
Tekniske data
Kemisk navn eller materiale Sulfo-N-succinimidyl 6-[3-(2-Pyridyldithio)propionamido] Hex
Mængde 50 mg
InChI formel InChI=1S/C18H23N3O8S3.Na/c22-14(8-11-30-31-15-6-3-5-10-20-15)19-9-4-1-2-7-17(24)29-21-16(23)12-13(18(21)25)32(26,27)28;/h3,5-6,10,13H,1-2,4,7-9,11-12H2,(H,19,22)(H,26,27,28);/q;+1/p-1
Formel vægt 527.0467