missing translation for 'onlineSavingsMsg'
Få mere at vide

Sulfiram, LoGiCal

Artikelnummer. 30459275
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459275 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30459275 Leverandør LoGiCal Leverandørnr. LGCFOR1945.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Sulfiram
CAS 95-05-6
Analyt- eller komponentnavne Sulfiram
Forsendelsestilstand Room Temperature
Molekylær formel C10 H20 N2 S3
Certificeringer/compliance ISO 9001
Synonym Disulfiram Imp. A (EP), Thiodicarbonic diamide ([(H2N)C(S)]2S), N,N,N',N'-tetraethyl-, Sulfide, bis(diethylthiocarbamoyl) (6CI,7CI,8CI), Thiodicarbonic diamide ([(H2N)C(S)]2S), tetraethyl- (9CI), Thiuram monosulfide, tetraethyl- (4CI), Bis(N,N-diethylthiocarbamoyl) sulfide, Bis(diethylthiocarbamoyl) sulfide, Bis(diethylthiocarbamyl) sulfide, Carbamodithioic acid, diethyl-, anhydrosulfide, Kutka, Kutkasin, Methanethioamide, 1,1'-thiobis[N,N-diethyl-, Monosulfiram, NSC 36731, Sanigal, Sarcocide B, Sulfide, bis[(diethylamino)thioxomethyl], Sulfiram, Sulfirame, Sulfiramum, TTMS, Tetmos, Tetmosol, Tetraethylthiuram monosulfide, Tetrucid, Disulfiram Impurity A
InChI formel InChI=1S/C10H20N2S3/c1-5-11(6-2)9(13)15-10(14)12(7-3)8-4/h5-8H2,1-4H3
Løsningstype Neat
SMIL CCN(CC)C(=S)SC(=S)N(CC)CC
IUPAC navn diethylcarbamothioyl N,N-diethylcarbamodithioate
Industry Type Clinical Testing
Molekylvægt (g/mol) 264.47
Mængde 10 mg
Formel vægt 264.0789 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.