Få mere at vide
Sulfathiazole Sodium Salt, TRC
Artikelnummer. 30006004
Skift visning
Mængde:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006004 10 g
30011750 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006004 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69910810G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 144-74-1
Molekylær formel C9 H8 N3 O2 S2 . Na
Molekylvægt (g/mol) 277.298
Synonym Benzenesulfonamide, 4-amino-N-2-thiazolyl-, monosodium salt (9CI), Sodium, (N1-2-thiazolylsulfanilamido)- (7CI), Sulfanilamide, N1-2-thiazolyl-, monosodium salt (8CI), Sulfathiazole, sodium deriv. (6CI), 2-(p-Amino-N-sodiobenzenesulfonamide)thiazole, 2-(p
IUPAC navn sodium;(4-aminophenyl)sulfonyl-(1,3-thiazol-2-yl)azanide
SMIL [Na+].Nc1ccc(cc1)S(=O)(=O)[N-]c2nccs2
Tekniske data
Kemisk navn eller materiale Sulfathiazole Sodium
Mængde 10 g
InChI formel InChI=1S/C9H8N3O2S2.Na/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9;/h1-6H,10H2;/q-1;+1
Formel vægt 276.996