Få mere at vide
Sulfate Standard: SO4(-2) @ 10000 μg/mL in H2O, VHG™
Artikelnummer. 17999714
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17999714 100 ml
17959154 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17999714 Leverandør VHG Labs Leverandørnr. VHGI1PSO4100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfate Standard: SO4(-2) @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfate
CAS 14808-79-8
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Sulfate
Forsendelsestilstand Room Temperature
Molekylær formel (S O4)2-
Certificeringer/compliance ISO 17034
Synonym SO42-
SMIL [O-]S(=O)(=O)[O-]
IUPAC navn Sulfate
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 96.06
Mængde 100 ml
Formel vægt 96.06 g/mol