Få mere at vide
Sulfapyridine-d4, TRC
Artikelnummer. 30416391
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30416391 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30416391 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6990811MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfapyridine-d4
CAS 1189863-86-2
Molekylær formel C11 2H4 H7 N3 O2 S
Mængde 1 mg
Synonym 4-Amino-N-2-pyridinyl-d4-benzenesulfonamide, 2-Sulfanilamidopyridine-d4, 2-Sulfapyridine-d4, 4-Amino-N-2-pyridinyl-d4-benzenesulfonamide, Adiplon-d4, Coccoclase-d4, Dagenan-d4, Eubasin-d4, Eubasinum-d4, Haptocil-d4, Sulfidine-d4, Sulphapyridine-d4, Thioseptal-d4, Trianon-d4
InChI formel InChI=1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14)/i1D,2D,3D,8D
SMIL [2H]c1nc(NS(=O)(=O)c2ccc(N)cc2)c([2H])c([2H])c1[2H]
IUPAC navn 4-amino-N-(3,4,5,6-tetradeuteriopyridin-2-yl)benzenesulfonamide
Molekylvægt (g/mol) 253.31
Formel vægt 253.0823