Få mere at vide
Sulfanilamide-d4 (ring-d4), TRC
Artikelnummer. 30425122
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30425122 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425122 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68910110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfanilamide-d4 (ring-d4)
CAS 77435-46-2
Molekylær formel C6 D4 H4 N2 O2 S
Mængde 10 mg
Synonym Benzene-2,3,5,6-d4-sulfonamide','4-amino- (9CI)
InChI formel InChI=1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)N
IUPAC navn 4-aminobenzene-2,3,5,6-d4-sulfonamide
Molekylvægt (g/mol) 176.2296
Formel vægt 176.0558