Få mere at vide
Sulfamonomethoxine Sodium, TRC
Artikelnummer. 30421342
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421342 10 mg
30415151 50 mg
30410215 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421342 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69927810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38006-08-5
Molekylær formel C11 H11 N4 O3 S . Na
Molekylvægt (g/mol) 302.28
Synonym Benzenesulfonamide, 4-amino-N-(6-methoxy-4-pyrimidinyl)-, monosodium salt (9CI), Sodium sulfamonomethoxine, Sulfamonomethoxine sodium, Sulfamonomethoxine sodium salt
IUPAC navn sodium;(4-aminophenyl)sulfonyl-(6-methoxypyrimidin-4-yl)azanide
SMIL [Na+].COc1cc([N-]S(=O)(=O)c2ccc(N)cc2)ncn1
Tekniske data
Kemisk navn eller materiale Sulfamonomethoxine sodium
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C11H11N4O3S.Na/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9;/h2-7H,12H2,1H3;/q-1;+1
Formel vægt 302.045
Procent renhed >95%