Få mere at vide
Sulfamethylphenazole, LoGiCal
Artikelnummer. 30459434
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459434 10 mg
1 options
Artikelnummer. 30459434 Leverandør LoGiCal Leverandørnr. LGCFOR1108.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfamethylphenazole
CAS 852-19-7
Analyt- eller komponentnavne Sulfamethylphenazole
Forsendelsestilstand Room Temperature
Molekylær formel C16 H16 N4 O2 S
Certificeringer/compliance ISO 9001
Synonym Sulfamethylphenazole, 4-Amino-N-(3-methyl-1-phenyl-1H-pyrazol-5-yl)benzenesulfonamide, N1-(3-Methyl-1-phenylpyrazol-5-yl)sulfanilamide, 1-Phenyl-3-methyl-5-sulfanilamidopyrazole, 3-(p-Aminobenzosulfoamido)-2-phenyl-5-methylpyrazole, 3-Methyl-1-phenyl-5-sulfanilamidopyrazole, 3-Sulfanilamido-5-methyl-2-phenylpyrazole, 5-(p-Aminobenzenesulfonamido)-1-phenyl-3-methylpyrazole, Ciba 18,605-Ba, N1-(3-Methyl-1-phenylpyrazol-5-yl)sulfanilamide, Sulfapyrazole, Sulfazamet, Vesulong
InChI formel InChI=1S/C16H16N4O2S/c1-12-11-16(20(18-12)14-5-3-2-4-6-14)19-23(21,22)15-9-7-13(17)8-10-15/h2-11,19H,17H2,1H3
Løsningstype Neat
SMIL Cc1cc(NS(=O)(=O)c2ccc(N)cc2)n(n1)c3ccccc3
IUPAC navn 4-amino-N-(5-methyl-2-phenylpyrazol-3-yl)benzenesulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 328.39
Mængde 10 mg
Formel vægt 328.0994 g/mol