Få mere at vide
Sulfamethoxazole-13C6, TRC
Artikelnummer. 17963965
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963965 10 mg
17910605 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17963965 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69908810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1196157-90-0
Molekylær formel 13C6 C4 H11 N3 O3 S
Molekylvægt (g/mol) 259.23
Synonym Benzene-1,2,3,4,5,6-13C6-sulfonamide, 4-amino-N-(5-methyl-3-isoxazolyl)-, 4-Amino-N-(5-methyl-3-isoxazolyl)benzene-1,2,3,4,5,6-13C6-sulfonamide
IUPAC navn 4-amino-N-(5-methyl-1,2-oxazol-3-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-sulfonamide
SMIL Cc1onc(NS(=O)(=O)[13c]2[13cH][13cH][13c](N)[13cH][13cH]2)c1
Tekniske data
Kemisk navn eller materiale Sulfamethoxazole ring-13C6
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C10H11N3O3S/c1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6H,11H2,1H3,(H,12,13)/i2+1,3+1,4+1,5+1,8+1,9+1
Formel vægt 259.0722
Procent renhed >95%