Få mere at vide
Sulfamethazine Related Compound A, TRC
Artikelnummer. 30415518
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30415518 1 mg -
30424421 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30415518 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6991181MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfaguanidine monohydrate
CAS 6190-55-2
Molekylær formel C7 H10 N4 O2 S . H2 O
Mængde 1 mg
Synonym Benzenesulfonamide','4-amino-N-(aminoiminomethyl)-','hydrate (1:1), Benzenesulfonamide','4-amino-N-(aminoiminomethyl)-','monohydrate (9CI), Sulfanilamide','N1-amidino-','monohydrate (8CI), N-p-Aminobenzenesulfonylguanidine monohydrate, Sulfaguanidine hydrate, Sulfaguanidine monohydrate, Sulphaguanidine monohydrate, p-Aminobenzenesulfonylguanidine monohydrate
InChI formel InChI=1S/C7H10N4O2S.H2O/c8-5-1-3-6(4-2-5)14(12,13)11-7(9)10;/h1-4H,8H2,(H4,9,10,11);1H2
SMIL O.NC(=N)NS(=O)(=O)c1ccc(N)cc1
IUPAC navn 1-(4-aminophenyl)sulfonylguanidine;hydrate
Molekylvægt (g/mol) 232.26
Formel vægt 232.063