Få mere at vide
Sulfameter-d4, TRC
Artikelnummer. 30412533
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412533 10 mg
30411444 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412533 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69904710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189483-96-2
Molekylær formel C11H8D4N4O3S
Molekylvægt (g/mol) 284.33
Synonym 4-Amino-N-(5-methoxy-2-pyrimidinyl)benzenesulfonamide-d4, 5-Methoxysulfadazine-d4, Sulfamethoxydiazine-d4, Sulfametin-d4, Sulfametorine-d4, Sulfamethoxine-d4, AHR-857-d4, Bayrena-d4, Durenat-d4, Kinecid-d4, Kiron-d4, Koroci-d4, Sulla-d4, Ultrax-d4
IUPAC navn 4-amino-2,3,5,6-tetradeuterio-N-(5-methoxypyrimidin-2-yl)benzenesulfonamide
SMIL NC1=C([2H])C([2H])=C(S(NC2=NC=C(OC)C=N2)(=O)=O)C([2H])=C1[2H]
Tekniske data
Kemisk navn eller materiale Sulfameter-d4
Mængde 10 mg
InChI formel InChI=1S/C11H12N4O3S/c1-18-9-6-13-11(14-7-9)15-19(16,17)10-4-2-8(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)/i2D,3D,4D,5D
Formel vægt 284.0881