Få mere at vide
Sulfadimethoxine, LoGiCal
Artikelnummer. 30462450
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462450 10 mg
1 options
Artikelnummer. 30462450 Leverandør LoGiCal Leverandørnr. LGCFOR3416.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfadimethoxine
CAS 122-11-2
Analyt- eller komponentnavne Sulfadimethoxine
Forsendelsestilstand Room Temperature
Molekylær formel C12 H14 N4 O4 S
Certificeringer/compliance ISO 9001
Synonym Benzenesulfonamide, 4-amino-N-(2,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(2,6-dimethoxy-4-pyrimidinyl)- (6CI,8CI), 2,4-Dimethoxy-6-sulfanilamido-1,3-diazine, 2,4-Dimethoxy-6-sulfanilamido-1,3-pyrimidine, 2,4-Dimethoxy-6-sulfanilamidopyrimidine, 2,4-Dimethoxy-6-sulfonylamido-1,3-diazine, 2,6-Dimethoxy-4-sulfanilamidopyrimidine, 4-(p-Aminobenzenesulfonamido)-2,6-dimethoxypyrimidine, 4-Sulfa-2,6-dimethoxypyrimidine, 4-Sulfanilamido-2,6-dimethoxypyrimidine, 6-Sulfanilamido-2,4-dimethoxy-1,3-diazine, 6-Sulfanilamido-2,4-dimethoxypyrimidine, Abcid, Agribon, Albon, Arnosulfan, Bactrovet, Deposul, Diasulfa, Dimethoxysulfadiazine, Dinosol, Dorisul, Fuxal, Lasibon, Madribon, Madrigid, Madriqid, Madroxin, Madroxine, Maxulvet, Mecozine, Metoxidon, N-(2,6-Dimethoxypyrimidin-4-yl)-4-aminobenzenesulfonamide, N1-(2,6-Dimethoxy-4-pyrimidinyl)sulfanilamide, NSC 683544, Neostreptal, Omnibon, Persulfen, Redifal, SDMO, Scandisil, Sudine, Suldixine, Sulfabon, Sulfadimethoxin, Sulfadimethoxine, Sulfadimethoxydiazine, Sulfadimetoxin, Sulfadimetoxine, Sulfadimoxine, Sulfamed G, Sulfastop, Sulfoplan, Sulforal, Sulphadimethoxine, Sulxin, Sumbio, Symbio, Ultrasulfon
InChI formel InChI=1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)
Løsningstype Neat
SMIL COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1
IUPAC navn 4-amino-N-(2,6-dimethoxypyrimidin-4-yl)benzenesulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 310.33
Mængde 10 mg
Formel vægt 310.0736 g/mol