Få mere at vide
Sulfadimethoxine 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sulfadimethoxine |
| CAS | 122-11-2 |
| Analyt- eller komponentnavne | Sulfadimethoxine |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C12 H14 N4 O4 S |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Benzenesulfonamide, 4-amino-N-(2,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(2,6-dimethoxy-4-pyrimidinyl)- (6CI,8CI), 2,4-Dimethoxy-6-sulfanilamido-1,3-diazine, 2,4-Dimethoxy-6-sulfanilamido-1,3-pyrimidine, 2,4-Dimethoxy-6-sulfanilamidopyrimidine, 2,4-Dimethoxy-6-sulfonylamido-1,3-diazine, 2,6-Dimethoxy-4-sulfanilamidopyrimidine, 4-(p-Aminobenzenesulfonamido)-2,6-dimethoxypyrimidine, 4-Sulfa-2,6-dimethoxypyrimidine, 4-Sulfanilamido-2,6-dimethoxypyrimidine, 6-Sulfanilamido-2,4-dimethoxy-1,3-diazine, 6-Sulfanilamido-2,4-dimethoxypyrimidine, Abcid, Agribon, Albon, Arnosulfan, Bactrovet, Deposul, Diasulfa, Dimethoxysulfadiazine, Dinosol, Dorisul, Fuxal, Lasibon, Madribon, Madrigid, Madriqid, Madroxin, Madroxine, Maxulvet, Mecozine, Metoxidon, N-(2,6-Dimethoxypyrimidin-4-yl)-4-aminobenzenesulfonamide, N1-(2,6-Dimethoxy-4-pyrimidinyl)sulfanilamide, NSC 683544, Neostreptal, Omnibon, Persulfen, Redifal, SDMO, Scandisil, Sudine, Suldixine, Sulfabon, Sulfadimethoxin, Sulfadimethoxine, Sulfadimethoxydiazine, Sulfadimetoxin, Sulfadimetoxine, Sulfadimoxine, Sulfamed G, Sulfastop, Sulfoplan, Sulforal, Sulphadimethoxine, Sulxin, Sumbio, Symbio, Ultrasulfon |
| InChI formel | InChI=1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16) |
| SMIL | COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1 |
| Vis mere |