Få mere at vide
Sulfachlorpyridazine-d4, TRC
Artikelnummer. 30418564
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30418564 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418564 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6890621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sulfachlorpyridazine-d4
CAS 1795037-54-5
Molekylær formel C10H5D4ClN4O2S
Mængde 1 mg
Synonym 4-Amino-N-(6-chloro-3-pyridazinyl)-benzenesulfonamide-d4, 3-Chloro-6-sulfanilamidopyridazine-d4, Sulfaclorazina-d4, Sulfarene-d4.
InChI formel InChI=1S/C10H9ClN4O2S/c11-9-5-6-10(14-13-9)15-18(16,17)8-3-1-7(12)2-4-8/h1-6H,12H2,(H,14,15)/i1D,2D,3D,4D
SMIL NC1=C([2H])C([2H])=C(S(NC2=NN=C(Cl)C=C2)(=O)=O)C([2H])=C1[2H]
IUPAC navn 4-amino-N-(6-chloropyridazin-3-yl)-2,3,5,6-tetradeuteriobenzenesulfonamide
Molekylvægt (g/mol) 288.75
Formel vægt 288.0386