missing translation for 'onlineSavingsMsg'
Få mere at vide

Sulfachloropyrazin, TRC

Artikelnummer. 30012255
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012255 1 g
17968504 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30012255 Supplier Toronto Research Chemicals Supplier No. TRCS6890631G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-65-8
Molekylær formel C10 H9 Cl N4 O2 S
Molekylvægt (g/mol) 284.72
Synonym Benzenesulfonamide, 4-amino-N-(6-chloropyrazinyl)- (9CI), Sulfanilamide, N1-(6-chloropyrazinyl)- (7CI,8CI), 4-Amino-N-(6-chloro-2-pyrazinyl)benzenesulfonamide, Esb3, Esb3 (antibiotic), N1-(6-Chloropyrazinyl)sulfanilamide, Sulfachloropyrazine, Sulfaclozine, Sulfatyf
IUPAC navn 4-amino-N-(6-chloropyrazin-2-yl)benzensulfonamid
SMIL Nc1ccc(cc1)S(=O)(=O)Nc2cncc(Cl)n2

Specifications

Kemisk navn eller materiale Sulfachloropyrazine
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C10H9ClN4O2S/c11-9-5-13-6-10(14-9)15-18(16,17)8-3-1-7(12)2-4-8/h1-6H,12H2,(H,14,15)
Formel vægt 284.0135
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.