missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUHW2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05966F
Additional Details : Weight : 0.00970kg
Description
SUHW2 Polyclonal specifically detects SUHW2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SUHW2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C19orf37, chromosome 19 open reading frame 37, Enzyme-like protein PIT13, MGC51082, Zfp428, zinc finger protein 428 | |
Produced in rabbits immunized with E. coli-derived Human SUHW2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
140883 | |
Store at 4°C in the dark. | |
IgG |