Learn More
Sudan III, C.I. 26100, Supelco™
Specifications
Specifications
| Melting Point | 199°C (Decomposition) |
| CAS | 85-86-9 |
| MDL Number | MFCD00003905 |
| Synonym | Sudan III, C.I. 26100, 1-(4-Phenylazophenylazo)-2-naphthol, Sudan III, C.I. 26100 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 352.39 |
| Grade | Research |