Få mere at vide
Sudan I-d5, TRC
Artikelnummer. 30414225
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30414225 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414225 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68888710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sudan 1 D5
CAS 752211-63-5
Molekylær formel C16 2H5 H7 N2 O
Mængde 10 mg
Synonym Sudan 1 D5 (phenyl D5), 2-Naphthalenol','1-(2-phenyl-2,3,4,5,6-d5-diazenyl)- (ACI), 1-(2-Phenyl-2,3,4,5,6-d5-diazenyl)-2-naphthalenol (ACI), 2-Naphthalenol','1-(phenyl-d5-azo)- (9CI)
InChI formel InChI=1S/C16H12N2O/c19-15-11-10-12-6-4-5-9-14(12)16(15)18-17-13-7-2-1-3-8-13/h1-11,19H/i1D,2D,3D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(N=Nc2c(O)ccc3ccccc23)c([2H])c1[2H]
IUPAC navn 1-[(E)-(2,3,4,5,6-pentadeuteriophenyl)diazenyl]naphthalen-2-ol
Molekylvægt (g/mol) 253.31
Formel vægt 253.1263