missing translation for 'onlineSavingsMsg'
Learn More

Sucrose Monocaprate (~90%), TRC

Product Code. 30421546
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30421546 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421546 Supplier Toronto Research Chemicals Supplier No. TRCS080893500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Sucrose Monocaprate
CAS 31835-06-0
Molekylær formel C22H40O12
Mængde 500 mg
Synonym Capric Acid Sucrose Monoester, Ryoto Sugar Ester MO-C 10, Ryoto Sugar Ester SM 1000, Saccharose Monocaprate, Sucrose Monocaprate, Sucrose Decanoate
InChI formel InChI=1S/C22H40O12/c1-2-3-4-5-6-7-8-9-15(25)31-11-14-16(26)18(28)19(29)21(32-14)34-22(12-24)20(30)17(27)13(10-23)33-22/h13-14,16-21,23-24,26-30H,2-12H2,1H3/t13-,14-,16-,17-,18+,19-,20+,21-,22+/m1/s1
SMIL O[C@H]1[C@H](O)[C@@](O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](COC(CCCCCCCCC)=O)O2)(CO)O[C@@H]1CO
IUPAC navn [(2R,3S,4S,5R,6R)-6-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methyl decanoate
Molekylvægt (g/mol) 496.55
Formel vægt 496.252
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.