Få mere at vide
Sucrose Brix/RI Standards, Paragon Scientific™
Artikelnummer. 16080410
Koncentration:
10,00 Brix
12,50 Brix
15,00 Brix
20,00 Brix
25,00 Brix
30,00 Brix
35 Brix
40,00 Brix
45,00 Brix
5,00 Brix
50,00 Brix
55 Brix
7,50 Brix
Pakningsstørrelse:
15ml
Artikelnummer. 16080410
Brand: Paragon Scientific SS25
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Primary level refractive index standards traceable to National Standards
  • Manufactured in accordance with ICUMSA Methods
  • Supplied in tamper evident packs with 6 weeks shelf life
Tekniske data
Koncentration 25,00 Brix
Mængde 15 mL
Angivne nominelle værdier