missing translation for 'onlineSavingsMsg'
Få mere at vide

Sucralfate (Technical Grade), TRC

Artikelnummer. 30426085
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426085 1 g
30414229 100 mg
30416708 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30426085 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6923501G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54182-58-0
Molekylær formel C12 H54 Al16 O75 S8
Molekylvægt (g/mol) 2086.74
Synonym Aluminum, hexadeca-μ-hydroxytetracosahydroxy[μ8-[[1,3,4,6-tetra-O-sulfo-β-D-fructofuranosyl α-D-glucopyranoside tetrakis(sulfato-κO')](8-)]]hexadeca-, Antepsin, Carafate, Citogel, Hexagastron, Keal, Succosa, Sucralafate, Sucralfate, Sucralfin, Sucrate, Sugast, Sulcrate, Ulcar, Ulcerlmin, Ulcermin, Ulcogant, Urbal, Venter Coordination Compound
SMIL O[Al+]1(O)[OH-][Al+](O)(OS(=O)(=O)OC[C@H]2O[C@H](O[C@]3(COS(=O)(=O)O[Al+]4(O)[OH-][Al+](O)(O)[OH-]4)O[C@H](COS(=O)(=O)O[Al+]5(O)[OH-][Al+](O)(O)[OH-]5)[C@@H](OS(=O)(=O)O[Al+]6(O)[OH-][Al+](O)(O)[OH-]6)[C@@H]3OS(=O)(=O)O[Al+]7(O)[OH-][Al+](O)(O)[OH-]7)[C@H](OS(=O)(=O)O[Al+]8(O)[OH-][Al+](O)(O)[OH-]8)[C@@H](OS(=O)(=O)O[Al+]9(O)[OH-][Al+](O)(O)[OH-]9)[C@@H]2OS(=O)(=O)O[Al+]%10(O)[OH-][Al+](O)(O)[OH-]%10)[OH-]1

Specifications

Kemisk navn eller materiale Sucralfate
Mængde 1 g
InChI formel InChI=1S/C12H22O35S8.16Al.40H2O/c13-48(14,15)37-1-4-6(43-51(22,23)24)8(45-53(28,29)30)9(46-54(31,32)33)11(40-4)42-12(3-39-50(19,20)21)10(47-55(34,35)36)7(44-52(25,26)27)5(41-12)2-38-49(16,17)18;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h4-11H,1-3H2,(H,13,14,15)(H,16,17,18)(H,19,20,21)(H,22,23,24)(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36);;;;;;;;;;;;;;;;;40*1H2/q;16*+3;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-48/t4-,5-,6-,7-,8+,9-,10+,11-,12+;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/m1......................................................../s1
Formel vægt 2085.5223
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.