Få mere at vide
Sucralfate (Technical Grade), TRC
Artikelnummer. 30414229
Skift visning
Mængde:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414229 100 mg
30426085 1 g
30416708 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30414229 Leverandør Toronto Research Chemicals Leverandørnr. TRCS692350100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54182-58-0
Molekylær formel C12 H54 Al16 O75 S8
Molekylvægt (g/mol) 2086.74
Synonym Aluminum, hexadeca-μ-hydroxytetracosahydroxy[μ8-[[1,3,4,6-tetra-O-sulfo-β-D-fructofuranosyl α-D-glucopyranoside tetrakis(sulfato-κO')](8-)]]hexadeca-, Antepsin, Carafate, Citogel, Hexagastron, Keal, Succosa, Sucralafate, Sucralfate, Sucralfin, Sucrate, Sugast, Sulcrate, Ulcar, Ulcerlmin, Ulcermin, Ulcogant, Urbal, Venter Coordination Compound
SMIL O[Al+]1(O)[OH-][Al+](O)(OS(=O)(=O)OC[C@H]2O[C@H](O[C@]3(COS(=O)(=O)O[Al+]4(O)[OH-][Al+](O)(O)[OH-]4)O[C@H](COS(=O)(=O)O[Al+]5(O)[OH-][Al+](O)(O)[OH-]5)[C@@H](OS(=O)(=O)O[Al+]6(O)[OH-][Al+](O)(O)[OH-]6)[C@@H]3OS(=O)(=O)O[Al+]7(O)[OH-][Al+](O)(O)[OH-]7)[C@H](OS(=O)(=O)O[Al+]8(O)[OH-][Al+](O)(O)[OH-]8)[C@@H](OS(=O)(=O)O[Al+]9(O)[OH-][Al+](O)(O)[OH-]9)[C@@H]2OS(=O)(=O)O[Al+]%10(O)[OH-][Al+](O)(O)[OH-]%10)[OH-]1
Tekniske data
Kemisk navn eller materiale Sucralfate
Mængde 100 mg
InChI formel InChI=1S/C12H22O35S8.16Al.40H2O/c13-48(14,15)37-1-4-6(43-51(22,23)24)8(45-53(28,29)30)9(46-54(31,32)33)11(40-4)42-12(3-39-50(19,20)21)10(47-55(34,35)36)7(44-52(25,26)27)5(41-12)2-38-49(16,17)18;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h4-11H,1-3H2,(H,13,14,15)(H,16,17,18)(H,19,20,21)(H,22,23,24)(H,25,26,27)(H,28,29,30)(H,31,32,33)(H,34,35,36);;;;;;;;;;;;;;;;;40*1H2/q;16*+3;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-48/t4-,5-,6-,7-,8+,9-,10+,11-,12+;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/m1......................................................../s1
Formel vægt 2085.5223