Learn More
Succinylsulfathiazole, TRC
Product Code. 30417553
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417553 50 g 50g
30420491 10 g -
30410275 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30417553 Supplier Toronto Research Chemicals Supplier No. TRCS69217050G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 116-43-8
Molecular Formula C13 H13 N3 O5 S2
Molecular Weight (g/mol) 355.39
Synonym Butanoic acid, 4-oxo-4-[[4-[(2-thiazolylamino)sulfonyl]phenyl]amino]-, Succinanilic acid, 4'-(2-thiazolylsulfamoyl)- (7CI,8CI), 2-(N4-Succinylsulfanilamido)thiazole, 4'-(2-Thiazolylsulfamoyl)succinanilic acid, Colistatin, Cremosuxidine, Kaoxidin, Kaoxidine, NSC 14193, NSC 163939, Rolsul, Succinylsulfathiazole, Succinylsulphathiazole, Sulfadigesin, Sulfasuccidin, Sulfasuccidine, Sulfasuccinil, Sulfasuccithiazole, Sulfasuxidine, Sulfenterone, Thiacyl, p-2-Thiazolylsulfamylsuccinanilic acid
IUPAC Name 4-oxo-4-[4-(1,3-thiazol-2-ylsulfamoyl)anilino]butanoic acid
SMILES OC(=O)CCC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2nccs2
Specifications
Quantity 50 g
InChI Formula InChI=1S/C13H13N3O5S2/c17-11(5-6-12(18)19)15-9-1-3-10(4-2-9)23(20,21)16-13-14-7-8-22-13/h1-4,7-8H,5-6H2,(H,14,16)(H,15,17)(H,18,19)
Formula Weight 355.0297
Chemical Name or Material Succinylsulfathiazole