Få mere at vide
Succinylacetone, TRC
Artikelnummer. 30010471
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010471 10 mg
30003667 100 mg
30005025 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010471 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69206010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51568-18-4
Molekylær formel C7 H10 O4
Molekylvægt (g/mol) 158.1519
IUPAC navn 4,6-dioxoheptanoic acid
SMIL CC(=O)CC(=O)CCC(=O)O
Tekniske data
Kemisk navn eller materiale Succinylacetone
Mængde 10 mg
InChI formel InChI=1S/C7H10O4/c1-5(8)4-6(9)2-3-7(10)11/h2-4H2,1H3,(H,10,11)
Formel vægt 158.0579